KEY424826530 Key Organics / BIONET

2-[7-(3-ethyl-1,2,4-oxadiazol-5-yl)-[1,2,4]triazolo[4,3-a]pyridin-3-yl]acetic acid

  • MDL number MFCD20528960
  • Cas Number : 1338682-10-2
  • Molecular Weight : 273.252

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433341

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3069_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart