KEY424826533 Key Organics / BIONET

3-(4-methylphenyl)-1,4,8-triazaspiro[4.5]dec-3-en-2-one

  • MDL number MFCD06751103
  • Cas Number : 887220-99-7
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433344

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3072_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart