KEY424826535 Key Organics / BIONET

3-[1-(3-fluorophenyl)-1H-1,2,3-triazol-4-yl]-1,2,4-oxadiazole-5-carbohydrazide

  • MDL number MFCD17780208
  • Cas Number : 1255783-18-6
  • Molecular Weight : 289.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433346

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3074_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart