KEY424826536 Key Organics / BIONET

2-phenyl-4H,5H-pyrazolo[1,5-a]pyrazine-4-thione

  • MDL number MFCD17780233
  • Cas Number : 933242-89-8
  • Molecular Weight : 227.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433347

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3075_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart