KEY424826537 Key Organics / BIONET

8-[3-(4-chlorophenyl)-1,2,4-oxadiazol-5-yl]-2H,3H-[1,2,4]triazolo[4,3-a]pyridin-3-one

  • MDL number MFCD14703518
  • Cas Number : 1239728-32-5
  • Molecular Weight : 313.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433348

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3076_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart