KEY424826540 Key Organics / BIONET

2-[4-(propan-2-yl)phenyl]-4H,5H-pyrazolo[1,5-a]pyrazin-4-one

  • MDL number MFCD25951875
  • Cas Number : 1338666-95-7
  • Molecular Weight : 253.305

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433351

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3079_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart