KEY424826541 Key Organics / BIONET

2-(3-fluorophenyl)-4H,5H-pyrazolo[1,5-a]pyrazin-4-one

  • MDL number MFCD25951877
  • Cas Number : 1338651-03-8
  • Molecular Weight : 229.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433352

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3080_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart