KEY424826546 Key Organics / BIONET

ethyl 3-{[4-(propan-2-yl)piperazin-1-yl]sulfonyl}-1H-pyrazole-4-carboxylate

  • MDL number MFCD18194693
  • Cas Number : 1260986-99-9
  • Molecular Weight : 330.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Green Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433357

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3086_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart