KEY424826548 Key Organics / BIONET

1-(3-chloro-4-fluorophenyl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD03834520
  • Cas Number : 696647-51-5
  • Molecular Weight : 257.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-chloro-4-fluorophenyl)-5-oxo-3-pyrrolidinecarboxylic acid
Inchi Code 1S/C11H9ClFNO3/c12-8-4-7(1-2-9(8)13)14-5-6(11(16)17)3-10(14)15/h1-2,4,6H,3,5H2,(H,16,17)
Inchi Key BMPMCEJUODXJTL-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433359

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3088_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart