KEY424826549 Key Organics / BIONET

1-(3-fluorophenyl)-3-methylurea

  • MDL number MFCD22192598
  • Cas Number : 38648-52-1
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433360

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3089_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart