KEY424826550 Key Organics / BIONET

3-phenyl-1,4,8-triazaspiro[4.5]dec-3-en-2-one

  • MDL number MFCD06751102
  • Cas Number : 887220-96-4
  • Molecular Weight : 229.283

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433361

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3090_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart