KEY424826552 Key Organics / BIONET

5-amino-4H-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD00179325
  • Cas Number : 3641-13-2
  • Molecular Weight : 128.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-amino-4H-1,2,4-triazole-3-carboxylic acid
Inchi Code 1S/C3H4N4O2/c4-3-5-1(2(8)9)6-7-3/h(H,8,9)(H3,4,5,6,7)
Inchi Key MVRGLMCHDCMPKD-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433363

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3092_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart