KEY424826555 Key Organics / BIONET

tert-butyl 4-(3-phenyl-1H-pyrazol-5-yl)piperazine-1-carboxylate

  • MDL number MFCD24850921
  • Molecular Weight : 328.416

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433366

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3095_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart