KEY424826557 Key Organics / BIONET

1-(4-fluorophenyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD07397776
  • Cas Number : 887035-89-4
  • Molecular Weight : 221.191

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433368

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3099_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart