KEY424826563 Key Organics / BIONET

1-{7H,8H,9H,10H-pyrazino[1,2-b]indazol-1-yl}piperazine hydrochloride

  • MDL number MFCD28023512
  • Cas Number : 1610376-97-0
  • Molecular Weight : 293.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433374

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3107_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart