KEY424826565 Key Organics / BIONET

4-(4-fluorophenyl)-2,2-dimethyl-2,5-dihydro-1H-imidazol-5-one

  • MDL number MFCD19104837
  • Cas Number : 899926-79-5
  • Molecular Weight : 206.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433376

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3110_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart