KEY424826571 Key Organics / BIONET

4,5-dimethyl 1-(2-oxopropyl)-1H-1,2,3-triazole-4,5-dicarboxylate

  • MDL number MFCD22549283
  • Cas Number : 1039766-69-2
  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433382

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3117_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart