KEY424826574 Key Organics / BIONET

1'H-spiro[cycloheptane-1,2'-quinazoline]-4'-thiol

  • MDL number MFCD28023513
  • Cas Number : 892299-03-5
  • Molecular Weight : 246.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433385

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3120_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart