KEY424826575 Key Organics / BIONET

1'H-spiro[cyclopentane-1,2'-quinazoline]-4'-thiol

  • MDL number MFCD28023514
  • Cas Number : 346458-41-1
  • Molecular Weight : 218.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433386

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3121_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart