KEY424826576 Key Organics / BIONET

6-fluoro-3-(4-methylpiperazine-1-carbonyl)quinolin-4-ol

  • MDL number MFCD16639697
  • Cas Number : 914658-84-7
  • Molecular Weight : 289.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433387

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3122_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart