KEY424826577 Key Organics / BIONET

1'H-spiro[cyclohexane-1,2'-quinazoline]-4'-thiol

  • MDL number MFCD28023515
  • Cas Number : 126492-23-7
  • Molecular Weight : 232.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433388

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3123_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart