KEY424826579 Key Organics / BIONET

ethyl 4-amino-3-phenyl-2-sulfanylidene-2,3-dihydro-1,3-thiazole-5-carboxylate

  • MDL number MFCD00858199
  • Cas Number : 7202-71-3
  • Molecular Weight : 280.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433390

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3127_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart