KEY424826580 Key Organics / BIONET

(Z)-N'-hydroxy-1,5-dimethyl-1H-pyrrole-2-carboximidamide

  • MDL number MFCD03756659
  • Cas Number : 500024-91-9
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Orange Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433391

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3128_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart