KEY424826585 Key Organics / BIONET

4-{6,8-dioxo-2H,5H,6H,7H,8H-[1,3]dioxolo[4,5-g]quinazolin-7-yl}butanoic acid

  • MDL number MFCD07655292
  • Cas Number : 892293-14-0
  • Molecular Weight : 292.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(6,8-dioxo-5,8-dihydro[1,3]dioxolo[4,5-g]quinazolin-7(6H)-yl)butanoic acid
Inchi Code 1S/C13H12N2O6/c16-11(17)2-1-3-15-12(18)7-4-9-10(21-6-20-9)5-8(7)14-13(15)19/h4-5H,1-3,6H2,(H,14,19)(H,16,17)
Inchi Key NFVMLPMTMHSYSH-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433396

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3133_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart