KEY424826586 Key Organics / BIONET

3-{6,8-dioxo-2H,5H,6H,7H,8H-[1,3]dioxolo[4,5-g]quinazolin-7-yl}propanoic acid

  • MDL number MFCD14726785
  • Cas Number : 892293-07-1
  • Molecular Weight : 278.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433397

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3134_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart