KEY424826588 Key Organics / BIONET

ethyl 5-amino-3-(3-chlorophenyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD28023516
  • Molecular Weight : 265.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433399

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3136_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart