KEY424826593 Key Organics / BIONET

2-(1H-imidazol-5-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD08060928
  • Cas Number : 65592-96-3
  • Molecular Weight : 184.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433404

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3141_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart