KEY424826600 Key Organics / BIONET

4-[3-(2-methoxyphenyl)-1,2,4-oxadiazol-5-yl]piperidine hydrochloride

  • MDL number MFCD27960949
  • Cas Number : 1018626-66-8
  • Molecular Weight : 295.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433411

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3148_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart