KEY424826603 Key Organics / BIONET

1-{[1-(2-methylphenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride

  • MDL number MFCD28023522
  • Cas Number : 1610377-13-3
  • Molecular Weight : 330.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-((1-(o-tolyl)-1H-1,2,3-triazol-5-yl)methyl)piperazine dihydrochloride
Inchi Code 1S/C14H19N5.2ClH/c1-12-4-2-3-5-14(12)19-13(10-16-17-19)11-18-8-6-15-7-9-18;;/h2-5,10,15H,6-9,11H2,1H3;2*1H
Inchi Key XSCWRNSORYXTGL-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433414

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3151_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart