KEY424826606 Key Organics / BIONET

1-{[1-(2-fluorophenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride

  • MDL number MFCD28023524
  • Cas Number : 1338692-39-9
  • Molecular Weight : 334.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433417

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3154_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart