KEY424826611 Key Organics / BIONET

4,5-diethyl 1-[2-(4-fluorophenyl)-2-oxoethyl]-1H-1,2,3-triazole-4,5-dicarboxylate

  • MDL number MFCD28023527
  • Cas Number : 1610376-99-2
  • Molecular Weight : 349.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433422

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3161_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart