KEY424826612 Key Organics / BIONET

4,5-dimethyl 1-[2-(4-fluorophenyl)-2-oxoethyl]-1H-1,2,3-triazole-4,5-dicarboxylate

  • MDL number MFCD28023528
  • Cas Number : 1610377-03-1
  • Molecular Weight : 321.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name dimethyl 1-(2-(4-fluorophenyl)-2-oxoethyl)-1H-1,2,3-triazole-4,5-dicarboxylate
Inchi Code 1S/C14H12FN3O5/c1-22-13(20)11-12(14(21)23-2)18(17-16-11)7-10(19)8-3-5-9(15)6-4-8/h3-6H,7H2,1-2H3
Inchi Key FFIREINYQKOBRZ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433423

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3162_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart