KEY424826613 Key Organics / BIONET

2-[3-(4-bromophenyl)-1,2,4-oxadiazol-5-yl]ethan-1-amine hydrochloride

  • MDL number MFCD28023529
  • Cas Number : 937666-44-9
  • Molecular Weight : 304.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433424

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3163_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart