KEY424826616 Key Organics / BIONET

1-[6-(2-fluorophenyl)pyridazin-3-yl]pyrrolidin-3-amine

  • MDL number MFCD28023531
  • Cas Number : 1610377-02-0
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433427

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3166_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart