KEY424826621 Key Organics / BIONET

tert-butyl 4-[(hydrazinecarbonyl)methyl]piperazine-1-carboxylate

  • MDL number MFCD06795935
  • Cas Number : 1338673-52-1
  • Molecular Weight : 258.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433432

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3171_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart