KEY424826624 Key Organics / BIONET

tert-butyl 4-{5H,6H,7H,8H,9H-[1,2,4]triazolo[4,3-a]azepin-3-yl}piperidine-1-carboxylate

  • MDL number MFCD28023532
  • Cas Number : 1610377-23-5
  • Molecular Weight : 320.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433435

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3177_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart