KEY424826625 Key Organics / BIONET

tert-butyl 2-{5H,6H,7H,8H,9H-[1,2,4]triazolo[4,3-a]azepin-3-yl}pyrrolidine-1-carboxylate

  • MDL number MFCD28023533
  • Cas Number : 1610376-95-8
  • Molecular Weight : 306.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433436

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3178_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart