KEY424826626 Key Organics / BIONET

3-{5H,6H,7H,8H,9H-[1,2,4]triazolo[4,3-a]azepin-3-yl}piperidine dihydrochloride

  • MDL number MFCD25975127
  • Cas Number : 1443423-29-7
  • Molecular Weight : 293.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433437

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3179_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart