KEY424826643 Key Organics / BIONET

2-(4-chlorophenyl)-5-(2-hydroxyethyl)-6-methyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD18915343
  • Cas Number : 82019-55-4
  • Molecular Weight : 264.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433454

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3206_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart