KEY424826646 Key Organics / BIONET

3-chloro-N-(5-chloro-2-hydroxyphenyl)propanamide

  • MDL number MFCD21294385
  • Cas Number : 1370592-24-7
  • Molecular Weight : 234.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433457

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3209_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart