KEY424826647 Key Organics / BIONET

2-methyl-11-oxo-11H-pyrido[2,1-b]quinazoline-6-carboxylic acid

  • MDL number MFCD20754075
  • Cas Number : 1111000-64-6
  • Molecular Weight : 254.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433458

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3210_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart