KEY424826648 Key Organics / BIONET

2-methyl-6-(trifluoromethyl)pyrimidin-4-ol

  • MDL number MFCD03265321
  • Cas Number : 2836-44-4
  • Molecular Weight : 178.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-6-(trifluoromethyl)-4-pyrimidinol
Inchi Code 1S/C6H5F3N2O/c1-3-10-4(6(7,8)9)2-5(12)11-3/h2H,1H3,(H,10,11,12)
Inchi Key DBDRDWKBAGNHHS-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/809207

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3211_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart