KEY424826651 Key Organics / BIONET

6,7-dimethoxy-2-oxo-1,2-dihydroquinoline-3-carbaldehyde

  • MDL number MFCD07506517
  • Cas Number : 101382-56-3
  • Molecular Weight : 233.223

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433462

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3214_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart