KEY424826654 Key Organics / BIONET

7-methoxy-3-{[(4-methoxyphenyl)amino]methyl}-1,2-dihydroquinolin-2-one

  • MDL number MFCD02572698
  • Cas Number : 335222-91-8
  • Molecular Weight : 310.353

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433465

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3217_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart