KEY424826661 Key Organics / BIONET

4-[3-(3,4-dimethylphenyl)-1,2,4-oxadiazol-5-yl]-1,2-dihydrophthalazin-1-one

  • MDL number MFCD19105023
  • Cas Number : 1325306-36-2
  • Molecular Weight : 318.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433472

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3227_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart