KEY424826664 Key Organics / BIONET

N-(4-fluorophenyl)-6-hydrazinylpyridine-3-sulfonamide

  • MDL number MFCD12444500
  • Cas Number : 1040059-76-4
  • Molecular Weight : 282.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433475

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3230_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart