KEY424826666 Key Organics / BIONET

4-ethyl-2-(1H-pyrrol-1-yl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD26130172
  • Cas Number : 1610377-11-1
  • Molecular Weight : 222.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433477

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3239_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart