KEY424826667 Key Organics / BIONET

4-propyl-2-(1H-pyrrol-1-yl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD14797777
  • Cas Number : 1031672-01-1
  • Molecular Weight : 236.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433478

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3240_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart