KEY424826680 Key Organics / BIONET

{5,7-dimethyl-[1,2,4]triazolo[1,5-a]pyrimidin-2-yl}methanol

  • MDL number MFCD11846959
  • Cas Number : 54535-00-1
  • Molecular Weight : 178.195

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433492

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4057_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart