KEY424826684 Key Organics / BIONET

3-(4-bromophenyl)-5-methyl-1,2,4-oxadiazole

  • MDL number MFCD00487341
  • Cas Number : 118183-92-9
  • Molecular Weight : 239.072

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433496

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4061_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart